missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human KIR2DL1/KIR2DS5 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 8 publications
£173.00 - £442.00
Specifications
| Antigen | KIR2DL1/KIR2DS5 |
|---|---|
| Clone | 143211 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Produktkode | Brand | Quantity | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Quantity | Pris | Antal & tilgængelighed | |||||
18738449
![]() |
R&D Systems
FAB1844P-025 |
25 Tests |
£173.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18787526
![]() |
R&D Systems
FAB1844P-100 |
100 Tests |
£442.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
KIR2DL1/KIR2DS5 Monoclonal specifically detects KIR2DL1/KIR2DS5 in Human samples. It is validated for Flow Cytometry.Tekniske data
| KIR2DL1/KIR2DS5 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD158a, CD158g, NKAT9 | |
| KIR2DL1 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human KIR2DL1/KIR2DS5 on transfected cells. It does not stain cells transfected with KIR2DL2, 2DL3, 2DL4, 2DL5, 2DS1, 2DS2, 2DS4, 3DL1, 3DL2, or 3DS1. |
| 143211 | |
| Flow Cytometry | |
| R-PE | |
| Mouse | |
| Human | |
| P43626 | |
| 3802 | |
| BaF3 mouse pro-B cell line transfected with human KIR2DL1/KIR2DS5 Accession # P43626 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel