missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human KIR2DL1/KIR2DS5 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 8 publications
Brand: R&D Systems FAB1844P-100
This item is not returnable.
View return policy
Description
KIR2DL1/KIR2DS5 Monoclonal specifically detects KIR2DL1/KIR2DS5 in Human samples. It is validated for Flow Cytometry.
Specifications
| KIR2DL1/KIR2DS5 | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD158a, CD158g, NKAT9 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3802 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 143211 | |
| Flow Cytometry 10 uL/10^6 cells | |
| P43626 | |
| KIR2DL1 | |
| BaF3 mouse pro-B cell line transfected with human KIR2DL1/KIR2DS5 Accession # P43626 | |
| 100 Tests | |
| Primary | |
| Detects human KIR2DL1/KIR2DS5 on transfected cells. It does not stain cells transfected with KIR2DL2, 2DL3, 2DL4, 2DL5, 2DS1, 2DS2, 2DS4, 3DL1, 3DL2, or 3DS1. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction