Learn More
IP3R-3 Mouse, Unlabeled, Clone: 2, BD
Mouse Monoclonal Antibody
£157.25 - £298.29
Specifications
| Antigen | IP3R-3 |
|---|---|
| Clone | 2 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15805399
|
BD Biosciences
610312 |
50 μg |
£164.00 £157.25 / 1 set Save £6.75 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15815399
|
BD Biosciences
610313 |
150 μg |
£311.00 £298.29 / 1 set Save £12.71 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Inositol 1,4,5-triphosphate (IP3) functions as a second messenger for many hormones, growth factors, and neurotransmitters. IP3 causes the release of Ca2+ from intracellular stores by binding specific receptors that are coupled to Ca2+ channels. A number of studies have identified a family of at least four IP3 receptors (IP3R). The type III receptor (IP3R-3) has been isolated and characterized in human and rat. IP3 receptors are commonly localized in the endoplasmic reticulum, but have also been identified in the nucleus and the plasma membrane. Co-expression of different IP3 receptors is detected in most tissues and cell lines. Although these receptors appear to have a similar specificity for inositol phosphates, the different receptors have been reported to have different affinities for IP3 as follows: type II > type I > type III.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
| IP3R-3 | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG2a | |
| Affinity Purified | |
| Inositol 1,4,5-triphosphate receptor type III |
| 2 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Bovine, Canine, Human, Mouse, Rat | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.