missing translation for 'onlineSavingsMsg'
Learn More

IP3R-3 Mouse, Unlabeled, Clone: 2, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610313

 View more versions of this product

Product Code. 15815399

  • ONLINE EXCLUSIVE PRICE
    £311.00
    £298.29 / 1 set
    Save £12.71
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Inositol 1,4,5-triphosphate (IP3) functions as a second messenger for many hormones, growth factors, and neurotransmitters. IP3 causes the release of Ca2+ from intracellular stores by binding specific receptors that are coupled to Ca2+ channels. A number of studies have identified a family of at least four IP3 receptors (IP3R). The type III receptor (IP3R-3) has been isolated and characterized in human and rat. IP3 receptors are commonly localized in the endoplasmic reticulum, but have also been identified in the nucleus and the plasma membrane. Co-expression of different IP3 receptors is detected in most tissues and cell lines. Although these receptors appear to have a similar specificity for inositol phosphates, the different receptors have been reported to have different affinities for IP3 as follows: type II > type I > type III.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

IP3R-3
Monoclonal
250μg/mL
Inositol 1,4,5-triphosphate receptor type III
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Western Blot
2
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
150 μg
Cell Biology
Bovine, Canine, Human, Mouse, Rat
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.