missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Integrin beta 1/CD29 PE-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
£242.00 - £333.00
Specifications
| Antigen | Integrin beta 1/CD29 |
|---|---|
| Clone | 265917 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18703639
![]() |
R&D Systems
FAB2405P-025 |
25 Tests |
£242.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18758539
![]() |
R&D Systems
FAB2405P-100 |
100 Tests |
£333.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Integrin beta 1/CD29 Monoclonal specifically detects Integrin beta 1/CD29 in Mouse samples. It is validated for Flow Cytometry.Specifications
| Integrin beta 1/CD29 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Supernatant | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD29, CD29 antigen, Fibronectin receptor subunit beta, FNRBVLAB, GPIIA, integrin beta-1, integrin VLA-4 beta subunit, integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includesMDF2, MSK12), ITGB1, MDF2, MSK12, very late activation protein, beta polypeptide, VLA-4 subunit beta, VLA-BETA | |
| ITGB1 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects mouse Integrin beta 1/CD29 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Integrin beta 1, recombinant mouse (rm) Integrin beta 2, rmIntegrin beta 3, rmIntegrin beta 4, or rmIntegrin beta 6 is observed. |
| 265917 | |
| Flow Cytometry | |
| R-PE | |
| Rat | |
| Mouse | |
| P09055 | |
| 3688 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin beta 1/CD29 Gln21-Asp728 Accession # P09055 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title