missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Integrin beta 1/CD29 PE-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems FAB2405P-100
This item is not returnable.
View return policy
Description
Integrin beta 1/CD29 Monoclonal specifically detects Integrin beta 1/CD29 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| Integrin beta 1/CD29 | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD29, CD29 antigen, Fibronectin receptor subunit beta, FNRBVLAB, GPIIA, integrin beta-1, integrin VLA-4 beta subunit, integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includesMDF2, MSK12), ITGB1, MDF2, MSK12, very late activation protein, beta polypeptide, VLA-4 subunit beta, VLA-BETA | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3688 | |
| Mouse | |
| Supernatant |
| Flow Cytometry | |
| 265917 | |
| Flow Cytometry 10 uL/10^6 cells | |
| P09055 | |
| ITGB1 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin beta 1/CD29 Gln21-Asp728 Accession # P09055 | |
| 100 Tests | |
| Primary | |
| Detects mouse Integrin beta 1/CD29 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Integrin beta 1, recombinant mouse (rm) Integrin beta 2, rmIntegrin beta 3, rmIntegrin beta 4, or rmIntegrin beta 6 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction