missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-13 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
£169.00 - £1790.00
Specifications
| Antigen | IL-13 |
|---|---|
| Clone | 31606 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Neutralization 1-4 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
| Applications | Western Blot, Neutralization, Flow Cytometry, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18142521
![]() |
R&D Systems
MAB2131-SP |
25 μg |
£169.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18701137
![]() |
R&D Systems
MAB2131-100 |
100 μg |
£447.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18130261
|
R&D Systems
MAB2131-500 |
500 μg |
£1790.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-13 Monoclonal specifically detects IL-13 in Human samples. It is validated for Western Blot, Neutralization, Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
| IL-13 | |
| LYOPH | |
| Western Blot, Neutralization, Flow Cytometry, CyTOF | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ALRHMGC116789, BHR1interleukin-13, IL13, IL-13MGC116788, interleukin 13, MGC116786, NC30, P600 | |
| IL13 | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 31606 | |
| Western Blot 1 ug/mL, Neutralization 1-4 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Human | |
| P35225 | |
| 3596 | |
| E. coli-derived recombinant human IL-13 Gly35-Asn146 Accession # P35225 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IL-13 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human IL-4 or recombinant mouse IL-13 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title