missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-13 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB2131-100
This item is not returnable.
View return policy
Description
IL-13 Monoclonal specifically detects IL-13 in Human samples. It is validated for Western Blot, Neutralization, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| IL-13 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Neutralization 1-4 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| P35225 | |
| IL13 | |
| E. coli-derived recombinant human IL-13 Gly35-Asn146 Accession # P35225 | |
| 100 μg | |
| Primary | |
| Detects human IL-13 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human IL-4 or recombinant mouse IL-13 is observed. | |
| Human | |
| Ascites |
| Western Blot, Neutralization, Flow Cytometry, CyTOF | |
| 31606 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ALRHMGC116789, BHR1interleukin-13, IL13, IL-13MGC116788, interleukin 13, MGC116786, NC30, P600 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 3596 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction