Learn More
IL-12 (p40/p70) Rat anti-Mouse, PE, Clone: C15.6, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 554479
Description
The C15.6 antibody reacts with both free and complexed (homodimer p80 and heterodimer p70) forms of the p40 subunit of mouse interleukin-12 (IL-12). The immunogen used to generate the C15.6 hybridoma was recombinant mouse IL-12 p70 protein. p40 has also been described as a subunit of IL-23 and thus it is possible that the C15.6 antibody will crossreact with IL-23.
Host Species: Rat
Clone: C15.6
Isotype: IgG1
Species Reactivity: Mouse
Immunogen: CHO-expressed recombinant mouse IL-12 p70 protein
Intracellular Staining
Specifications
| IL-12 (p40/p70) | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| CHO-expressed recombinant mouse IL-12 p70 protein | |
| 0.1 mg | |
| Immunology | |
| Mouse | |
| IgG1 |
| Flow Cytometry | |
| C15.6 | |
| PE | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.