Learn More
IL-12 (p40/p70) Rat anti-Mouse, PE, Clone: C15.6, BD
Rat Monoclonal Antibody
£81.25 - £324.99
Specifications
| Antigen | IL-12 (p40/p70) |
|---|---|
| Clone | C15.6 |
| Concentration | 0.2mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15811589
|
BD Biosciences
562038 |
25 μg |
£84.55 £81.25 / Each Save £3.30 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15817728
|
BD Biosciences
554479 |
0.1 mg |
£339.00 £324.99 / Each Save £14.01 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The C15.6 monoclonal antibody specifically binds to both free and complexed (homodimer p80 and heterodimer p70) forms of the p40 subunit of mouse interleukin-12 (IL-12). The immunogen used to generate the C15.6 hybridoma was recombinant mouse IL-12 p70 protein. p40 has also been described as a subunit of IL-23 and thus it is possible that the C15.6 antibody will crossreact with IL-23.
Specifications
| IL-12 (p40/p70) | |
| 0.2mg/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| CHO-expressed recombinant mouse IL-12 p70 protein | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| C15.6 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.