missing translation for 'onlineSavingsMsg'
Learn More

anti-IgG1, κ, Mouse, Alexa Fluor™ 647, Clone: MOPC-21, BD

£269.49 - £308.82

Specifications

Clone MOPC-21
Applications Flow Cytometry, Western Blot
Conjugate Alexa Fluor 647
Host Species Mouse
Target Species Human
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15859048
View Documents
BD Biosciences
557783
50 Tests
ONLINE EXCLUSIVE PRICE
£471.00
£269.49 / Each
Save £201.51
42% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15879018
View Documents
BD Biosciences
557732
100 Tests
ONLINE EXCLUSIVE PRICE
£312.00
£308.82 / Each
Save £3.18
1% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The MOPC-21 immunoglobulin is a mouse myeloma protein. The MOPC-21 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues.

Host Species: Mouse
Clone: MOPC-21
Isotype: IgG1 κ
Species Reactivity: Human

Flow Cytometry, Intracellular Staining, Isotype Control

Specifications

Specifications

MOPC-21
Alexa Fluor 647
Human
Affinity Purified
Flow Cytometry, Western Blot
Mouse
IgG1 κ
Undiluted at 4°C
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.