Learn More
anti-IgG1, κ, Mouse, Alexa Fluor™ 647, Clone: MOPC-21, BD
Brand: BD Biosciences 557732
Description
The MOPC-21 immunoglobulin is a mouse myeloma protein. The MOPC-21 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues.
Host Species: Mouse
Clone: MOPC-21
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry, Intracellular Staining, Isotype Control
Specifications
| Flow Cytometry, Western Blot | |
| Alexa Fluor 647 | |
| Affinity Purified | |
| Human | |
| IgG1 κ |
| MOPC-21 | |
| Mouse | |
| 100 Tests | |
| Undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.