missing translation for 'onlineSavingsMsg'
Learn More

I-A/I-E Rat anti-Mouse, FITC, Clone: 2G9, BD

Rat Monoclonal Antibody

Brand:  BD Biosciences 562009

 View more versions of this product

Product Code. 15821559

  • ONLINE EXCLUSIVE PRICE
    £90.95
    £87.38 / Each
    Save £3.57
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The 2G9 antibody reacts with the I-Ad and I-Ed MHC class II alloantigens. It also reacts with transfectants expressing I-Eb and I-Ek and with cells from mice of the H-2p and H-2q haplotypes. This antibody is non-reactive with cells from SJL (H-2s) and NOD (H-2g7) mice. Flow cytometric analysis indicates that the 2G9 and M5/114.15.2 (Cat. No. 556999, for purified mAb) monoclonal antibodies have comparable reactivity on cells from mice with I-Ab, I-Ad, I-Aq, I-Ed, and I-Ek alloantigens.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

I-A/I-E
Monoclonal
0.5mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
BALB/c mouse epidermal Langerhans cells
50 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
2G9
FITC
Rat
Affinity Purified
RUO
Mouse
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.