missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Human NCAM1/CD56 (B-A19), Proteintech
Mouse Monoclonal Antibody
£49.00 - £151.00
Specifications
| Antigen | NCAM1/CD56 |
|---|---|
| Clone | B-A19 |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30233484
|
Proteintech
65264-1-IG-100UG |
100 μg |
£52.00 £49.00 / 100µg Save £3.00 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30237654
|
Proteintech
65264-1-IG-500UG |
500 μg |
£163.00 £151.00 / 500µg Save £12.00 7% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NCAM1/CD56 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human NCAM1/CD56. This antibody recognizes human antigen. The NCAM1/CD56 antibody has been validated for the following applications: FC analysis.Specifications
| NCAM1/CD56 | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 4684 | |
| IgG1 | |
| Affinity purification | |
| Flow Antibody |
| B-A19 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| CD56; MSK39; NCAM | |
| Primary | |
| Store at 2-8°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title