missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Human NCAM1/CD56 (B-A19), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 65264-1-IG-100UG
This item is not returnable.
View return policy
Description
The NCAM1/CD56 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human NCAM1/CD56. This antibody recognizes human antigen. The NCAM1/CD56 antibody has been validated for the following applications: FC analysis.Specifications
| NCAM1/CD56 | |
| Monoclonal | |
| 0.5 mg/mL | |
| CD56; MSK39; NCAM | |
| Affinity purification | |
| RUO | |
| 4684 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |
| Flow Cytometry | |
| B-A19 | |
| Unconjugated | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Flow Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction