missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Human DPP4/CD26 (BA5b), Proteintech
Mouse Monoclonal Antibody
£45.00 - £134.00
Specifications
| Antigen | DPP4/CD26 |
|---|---|
| Clone | BA5b |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235934
|
Proteintech
65267-1-IG-500UG |
500 μg |
£138.00 £134.00 / 500µg Save £4.00 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30234819
|
Proteintech
65267-1-IG-100UG |
100 μg |
£45.75 £45.00 / 100µg Save £0.75 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The DPP4/CD26 antibody from Proteintech is a mouse monoclonal antibody generated with a human t cell clone of human DPP4/CD26. This antibody recognizes human antigen. The DPP4/CD26 antibody has been validated for the following applications: FC analysis.Specifications
| DPP4/CD26 | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 1803 | |
| A human T cell clone | |
| Primary | |
| Store at 2-8°C. Stable for one year after shipment. |
| BA5b | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| ADABP; ADCP2; CD26; DPPIV; TP103 | |
| IgG2a κ | |
| Affinity purification | |
| Flow Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title