Learn More
Anti-Human DPP4/CD26 (BA5b), Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 65267-1-IG-500UG
 View more versions of this product
Product Code. 30235934
  • ONLINE EXCLUSIVE PRICE
    £138.00
    £134.00 / 500µg
    Save £4.00
    2% Off
Description
The DPP4/CD26 antibody from Proteintech is a mouse monoclonal antibody generated with a human t cell clone of human DPP4/CD26. This antibody recognizes human antigen. The DPP4/CD26 antibody has been validated for the following applications: FC analysis.
Specifications
Specifications
DPP4/CD26
Monoclonal
0.5 mg/mL
ADABP; ADCP2; CD26; DPPIV; TP103
A human T cell clone
500 μg
Primary
Human
Flow Antibody
IgG2a κ
Flow Cytometry
BA5b
Unconjugated
Mouse
Affinity purification
RUO
1803
Store at 2-8°C. Stable for one year after shipment.