missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Human CD81 (5A6), Proteintech
Mouse Monoclonal Antibody
£57.00 - £177.00
Specifications
| Antigen | CD81 |
|---|---|
| Clone | 5A6 |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30236413
|
Proteintech
65195-1-IG-500UG |
500 μg |
£188.00 £177.00 / 500µg Save £11.00 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30233737
|
Proteintech
65195-1-IG-100UG |
100 μg |
£60.00 £57.00 / 100µg Save £3.00 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD81 antibody from Proteintech is a mouse monoclonal antibody generated with human oci-ly8 cell line of human CD81. This antibody recognizes human antigen. The CD81 antibody has been validated for the following applications: FC analysis.Specifications
| CD81 | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 975 | |
| Human OCI-LY8 cell line | |
| Primary | |
| Store at 2-8°C. Stable for one year after shipment. |
| 5A6 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| S5.7; TAPA1; TSPAN28 | |
| IgG1 κ | |
| Protein A purification | |
| Flow Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title