Learn More
Anti-Human CD81 (5A6), Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 65195-1-IG-100UG
 View more versions of this product
Product Code. 30233737
  • ONLINE EXCLUSIVE PRICE
    £60.00
    £57.00 / 100µg
    Save £3.00
    5% Off
Description
The CD81 antibody from Proteintech is a mouse monoclonal antibody generated with human oci-ly8 cell line of human CD81. This antibody recognizes human antigen. The CD81 antibody has been validated for the following applications: FC analysis.
Specifications
Specifications
CD81
Monoclonal
0.5 mg/mL
S5.7; TAPA1; TSPAN28
Human OCI-LY8 cell line
100 μg
Primary
Human
Flow Antibody
IgG1 κ
Flow Cytometry
5A6
Unconjugated
Mouse
Protein A purification
RUO
975
Store at 2-8°C. Stable for one year after shipment.