missing translation for 'onlineSavingsMsg'
Learn More

anti-Human CD62L; Clone: DREG-56; BD

Brand:  BD Biosciences 562720

 View more versions of this product

Product Code. 15852179

  • ONLINE EXCLUSIVE PRICE
    £189.00
    £186.61 / Each
    Save £2.39
    1% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Host Species: Mouse
Clone: DREG-56
Isotype: IgG1, κ
Cross Reactivity Species: Hu
Immunogen: Supernatant from PMA-activated Human Peripheral Blood Leukocytes

Applications:

Flow Cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Human CD62L
DREG-56
Mouse
Affinity Purified
RUO
Human
Monoclonal
Brilliant Violet 605
Supernatant from PMA-activated Human Peripheral Blood Leukocytes
25 Tests
Primary
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.