missing translation for 'onlineSavingsMsg'
Learn More

anti-Human CD62L; Clone: DREG-56; BD

£186.61 - £361.57

Specifications

Antigen Human CD62L
Clone DREG-56
Classification Monoclonal
Conjugate Brilliant Violet 605
Host Species Mouse
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15852179
View Documents
BD Biosciences
562720
25 Tests
ONLINE EXCLUSIVE PRICE
£189.00
£186.61 / Each
Save £2.39
1% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15842179
View Documents
BD Biosciences
562719
100 Tests
ONLINE EXCLUSIVE PRICE
£369.00
£361.57 / Each
Save £7.43
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Host Species: Mouse
Clone: DREG-56
Isotype: IgG1, κ
Cross Reactivity Species: Hu
Immunogen: Supernatant from PMA-activated Human Peripheral Blood Leukocytes

Applications:

Flow Cytometry

Specifications

Specifications

Human CD62L
Monoclonal
Mouse
Human
Primary
DREG-56
Brilliant Violet 605
RUO
Supernatant from PMA-activated Human Peripheral Blood Leukocytes
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.