missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-Human CD62L; Clone: DREG-56; BD
Brand: BD Biosciences 562719
This item is not returnable.
View return policy
Description
Host Species: Mouse
Clone: DREG-56
Isotype: IgG1, κ
Cross Reactivity Species: Hu
Immunogen: Supernatant from PMA-activated Human Peripheral Blood Leukocytes
Applications:
Flow Cytometry
Specifications
| Human CD62L | |
| DREG-56 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human |
| Monoclonal | |
| Brilliant Violet 605 | |
| Supernatant from PMA-activated Human Peripheral Blood Leukocytes | |
| 100 Tests | |
| Primary |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction