missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Human CD58 (TS2/9), Proteintech
Mouse Monoclonal Antibody
£49.00 - £151.00
Specifications
| Antigen | CD58 |
|---|---|
| Clone | TS2/9 |
| Concentration | 500 μg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30233416
|
Proteintech
65193-1-IG-500UG |
500 μg |
£163.00 £151.00 / 500µg Save £12.00 7% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30238042
|
Proteintech
65193-1-IG-100UG |
100 μg |
£52.00 £49.00 / 100µg Save £3.00 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD58 antibody from Proteintech is a mouse monoclonal antibody generated with human cytolytic t cells of human CD58. This antibody recognizes human antigen. The CD58 antibody has been validated for the following applications: FC analysis.Specifications
| CD58 | |
| 500 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 965 | |
| Human cytolytic T cells | |
| Primary | |
| Store at 2-8°C. Stable for one year after shipment. |
| TS2/9 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| FLJ23181; FLJ43722; LFA-3; LFA3 | |
| IgG1 κ | |
| Protein G purification | |
| Flow Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title