missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Human CD58 (TS2/9), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 65193-1-IG-500UG
This item is not returnable.
View return policy
Description
The CD58 antibody from Proteintech is a mouse monoclonal antibody generated with human cytolytic t cells of human CD58. This antibody recognizes human antigen. The CD58 antibody has been validated for the following applications: FC analysis.Specifications
| CD58 | |
| Monoclonal | |
| 500 μg/mL | |
| FLJ23181; FLJ43722; LFA-3; LFA3 | |
| Human cytolytic T cells | |
| 500 μg | |
| Primary | |
| Human | |
| Flow Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| TS2/9 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 965 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction