missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Human CD55 (JS11), Proteintech
Mouse Monoclonal Antibody
£52.00 - £178.00
Specifications
| Antigen | CD55 |
|---|---|
| Clone | JS11 |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30237815
|
Proteintech
65271-1-IG-500UG |
500 μg |
£181.00 £178.00 / 500µg Save £3.00 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30233821
|
Proteintech
65271-1-IG-100UG |
100 μg |
£52.85 £52.00 / 100µg Save £0.85 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD55 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD55. This antibody recognizes human antigen. The CD55 antibody has been validated for the following applications: FC analysis.Specifications
| CD55 | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 1604 | |
| IgG1 | |
| Affinity purification | |
| Flow Antibody |
| JS11 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| CR; CROM; DAF; TC | |
| Primary | |
| Store at 2-8°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title