Learn More
Anti-Human CD55 (JS11), Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 65271-1-IG-500UG
 View more versions of this product
Product Code. 30237815
  • ONLINE EXCLUSIVE PRICE
    £181.00
    £178.00 / 500µg
    Save £3.00
    1% Off
Description
The CD55 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD55. This antibody recognizes human antigen. The CD55 antibody has been validated for the following applications: FC analysis.
Specifications
Specifications
CD55
Monoclonal
0.5 mg/mL
CR; CROM; DAF; TC
Affinity purification
RUO
1604
Store at 2-8°C. Stable for one year after shipment.
Liquid
Flow Cytometry
JS11
Unconjugated
Mouse
500 μg
Primary
Human
Flow Antibody
IgG1