missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Human CD31 (WM-59), Proteintech
Mouse Monoclonal Antibody
£46.00 - £158.00
Specifications
| Antigen | CD31 |
|---|---|
| Clone | WM-59 |
| Concentration | 0.5 mg/mL |
| Applications | Immunocytochemistry/Immunofluorescence, Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30237589
|
Proteintech
65268-1-IG-100UG |
100 μg |
£46.75 £46.00 / 100µg Save £0.75 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30237639
|
Proteintech
65268-1-IG-500UG |
500 μg |
£161.00 £158.00 / 500µg Save £3.00 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD31 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD31. This antibody recognizes human antigen. The CD31 antibody has been validated for the following applications: IF/ICC, FC analysis.Specifications
| CD31 | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 5175 | |
| IgG1 κ | |
| Affinity purification | |
| Flow Antibody |
| WM-59 | |
| Immunocytochemistry/Immunofluorescence, Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| CD31; FLJ58394; PECAM-1 | |
| Primary | |
| Store at 2-8°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title