missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Human CD31 (WM-59), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 65268-1-IG-100UG
This item is not returnable.
View return policy
Description
The CD31 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD31. This antibody recognizes human antigen. The CD31 antibody has been validated for the following applications: IF/ICC, FC analysis.Specifications
| CD31 | |
| Monoclonal | |
| 0.5 mg/mL | |
| CD31; FLJ58394; PECAM-1 | |
| Affinity purification | |
| RUO | |
| 5175 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |
| Immunocytochemistry/Immunofluorescence, Flow Cytometry | |
| WM-59 | |
| Unconjugated | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Flow Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction