missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Human CD163 (GHI/61), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 65169-1-IG-100UG
This item is not returnable.
View return policy
Description
The CD163 antibody from Proteintech is a mouse monoclonal antibody generated with glycoprotein preparation from hairy cell leukemia spleen of human CD163. This antibody recognizes human antigen. The CD163 antibody has been validated for the following applications: FC analysis.Specifications
| CD163 | |
| Monoclonal | |
| 0.5 mg/mL | |
| M130; MM130 | |
| Glycoprotein preparation from hairy cell leukemia spleen | |
| 100 μg | |
| Primary | |
| Human | |
| Flow Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| GHI/61 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 9332 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction