missing translation for 'onlineSavingsMsg'
Learn More

Anti-Human CD163 (GHI/61), Proteintech

Mouse Monoclonal Antibody

£49.00 - £147.00

Specifications

Antigen CD163
Clone GHI/61
Concentration 0.5 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30235767
View Documents
Proteintech
65169-1-IG-100UG
100 μg
ONLINE EXCLUSIVE PRICE
£52.00
£49.00 / 100µg
Save £3.00
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30237961
View Documents
Proteintech
65169-1-IG-500UG
500 μg
ONLINE EXCLUSIVE PRICE
£163.00
£147.00 / 500µg
Save £16.00
9% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The CD163 antibody from Proteintech is a mouse monoclonal antibody generated with glycoprotein preparation from hairy cell leukemia spleen of human CD163. This antibody recognizes human antigen. The CD163 antibody has been validated for the following applications: FC analysis.
Specifications

Specifications

CD163
0.5 mg/mL
Monoclonal
Liquid
RUO
9332
Glycoprotein preparation from hairy cell leukemia spleen
Primary
Store at 2-8°C. Stable for one year after shipment.
GHI/61
Flow Cytometry
Unconjugated
Mouse
Human
M130; MM130
IgG1 κ
Protein A purification
Flow Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.