missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Human ALCAM (3A6), Proteintech
Mouse Monoclonal Antibody
£60.00 - £184.00
Specifications
| Antigen | ALCAM |
|---|---|
| Clone | 3A6 |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30234906
|
Proteintech
65223-1-IG-500UG |
500 μg |
£187.00 £184.00 / 500µg Save £3.00 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236422
|
Proteintech
65223-1-IG-100UG |
100 μg |
£61.00 £60.00 / 100µg Save £1.00 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ALCAM antibody from Proteintech is a mouse monoclonal antibody generated with cultured human thymic epithelial cells of human ALCAM. This antibody recognizes human antigen. The ALCAM antibody has been validated for the following applications: FC analysis.Specifications
| ALCAM | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 214 | |
| Cultured human thymic epithelial cells | |
| Primary | |
| Store at 2-8°C. Stable for one year after shipment. |
| 3A6 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| CD166; FLJ38514; MEMD; MGC71733 | |
| IgG1 κ | |
| Affinity purification | |
| Flow Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title