missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Human ALCAM (3A6), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 65223-1-IG-500UG
This item is not returnable.
View return policy
Description
The ALCAM antibody from Proteintech is a mouse monoclonal antibody generated with cultured human thymic epithelial cells of human ALCAM. This antibody recognizes human antigen. The ALCAM antibody has been validated for the following applications: FC analysis.Specifications
| ALCAM | |
| Monoclonal | |
| 0.5 mg/mL | |
| CD166; FLJ38514; MEMD; MGC71733 | |
| Cultured human thymic epithelial cells | |
| 500 μg | |
| Primary | |
| Human | |
| Flow Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| 3A6 | |
| Unconjugated | |
| Mouse | |
| Affinity purification | |
| RUO | |
| 214 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction