missing translation for 'onlineSavingsMsg'
Learn More

HLA-A,B,C Mouse anti-Human, FITC, Clone: G46-2.6, BD

Mouse Monoclonal Antibody

£203.30 - £225.78

Specifications

Antigen HLA-A,B,C
Clone G46-2.6
Applications Flow Cytometry
Classification Monoclonal
Conjugate FITC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15838888
View Documents
BD Biosciences
557348
50 Tests
ONLINE EXCLUSIVE PRICE
£206.00
£203.30 / Each
Save £2.70
1% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15878258
View Documents
BD Biosciences
555552
100 Tests
ONLINE EXCLUSIVE PRICE
£229.00
£225.78 / Each
Save £3.22
1% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Reacts with a monomorphic epitope of major histocompatibility Class I antigens widely distributed on human nucleated cells. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: G46-2.6
Isotype: IgG1 κ
Species Reactivity: Human, Baboon/Rhesus/Cynomolgus

Flow Cytometry

Specifications

Specifications

HLA-A,B,C
Flow Cytometry
FITC
RUO
Baboon, Cynomolgus, Human, Rhesus Monkey
IgG1 κ
Affinity Purified
G46-2.6
Monoclonal
Mouse
Immunology
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.