Learn More
HLA-A,B,C Mouse anti-Human, FITC, Clone: G46-2.6, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 557348
Description
Reacts with a monomorphic epitope of major histocompatibility Class I antigens widely distributed on human nucleated cells. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: G46-2.6
Isotype: IgG1 κ
Species Reactivity: Human, Baboon/Rhesus/Cynomolgus
Flow Cytometry
Specifications
| HLA-A,B,C | |
| Monoclonal | |
| FITC | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| G46-2.6 | |
| Major histocompatibility complex, class I, A,B,C, HLA class I A,B,C | |
| Mouse | |
| 50 Tests | |
| Immunology | |
| Baboon, Cynomolgus, Human, Rhesus Monkey | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.