missing translation for 'onlineSavingsMsg'
Learn More

gp91[phox] Mouse, Unlabeled, Clone: 53, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 611414

 View more versions of this product

Product Code. 15895919

  • £171.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Although dormant in resting granulocytes, macrophages, and B lymphocytes, the neutrophil respiratory burst oxidase (NADPH-oxidase) generates superoxide and secondary oxygen-derived toxic products in response to bacteria or a variety of soluble stimuli. It is an integral membrane cytochrome, b558, which consists of two subunits, gp91[phox] and p21[phox]. Upon stimulation, cytochrome b558 forms a complex with cytosolic proteins, p67[phox], p47[phox], p40[phox], and rac2 and produces superoxide anions in a NADPH-dependent
manner. In chronic granulomatous disease (CGD), severe recurrent bacterial and fungal infections result from defective NADPH-oxidase activity. The majority of CGD cases are caused by a defective gp91[phox] gene. gp91[phox], implicated as a docking site for p47[phox], is membrane glycoprotein with multiple N-terminal hydrophobic domains and a hydrophilic C-terminus. The expression of gp91[phox] is restricted to terminally differentiated phagocytes and B lymphocytes. This cell type- and developmental stage-specific expression may be
controlled by transcriptional activators, such as PU.1 and YY1, and transcriptional repressors, such as CCAAT displacement protein (CDP). Endogenous mouse gp91phox shows 58kDa band, instead of 91kDa observed in human. Both mouse and human deglycosylated gp91 are 54kDa. This difference may be due to less glycosylation sites in the mouse sequence.

Host Species: Mouse
Clone: 53
Isotype: IgG1
Species Reactivity: Mouse
Immunogen: Mouse gp91[phox] aa. 450-556
Formula Weight [Chemical]: 58kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

gp91[phox]
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Mouse gp91[phox] aa. 450-556
50 μg
Cell Biology
Mouse, Rat
IgG1
Western Blot
53
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.