Learn More
gp91[phox] Mouse, Unlabeled, Clone: 53, BD
Mouse Monoclonal Antibody
£267.00 - £314.49
Specifications
| Antigen | gp91[phox] |
|---|---|
| Clone | 53 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15895919
|
BD Biosciences
611414 |
50 μg |
£267.00
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15805929
|
BD Biosciences
611415 |
150 μg |
£328.00 £314.49 / 1 set Save £13.51 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Although dormant in resting granulocytes, macrophages, and B lymphocytes, the neutrophil respiratory burst oxidase (NADPH-oxidase) generates superoxide and secondary oxygen-derived toxic products in response to bacteria or a variety of soluble stimuli. It is an integral membrane cytochrome, b558, which consists of two subunits, gp91[phox] and p21[phox]. Upon stimulation, cytochrome b558 forms a complex with cytosolic proteins, p67[phox], p47[phox], p40[phox], and rac2 and produces superoxide anions in a NADPH-dependent
manner. In chronic granulomatous disease (CGD), severe recurrent bacterial and fungal infections result from defective NADPH-oxidase activity. The majority of CGD cases are caused by a defective gp91[phox] gene. gp91[phox], implicated as a docking site for p47[phox], is membrane glycoprotein with multiple N-terminal hydrophobic domains and a hydrophilic C-terminus. The expression of gp91[phox] is restricted to terminally differentiated phagocytes and B lymphocytes. This cell type- and developmental stage-specific expression may be
controlled by transcriptional activators, such as PU.1 and YY1, and transcriptional repressors, such as CCAAT displacement protein (CDP). Endogenous mouse gp91phox shows 58kDa band, instead of 91kDa observed in human. Both mouse and human deglycosylated gp91 are 54kDa. This difference may be due to less glycosylation sites in the mouse sequence.
Host Species: Mouse
Clone: 53
Isotype: IgG1
Species Reactivity: Mouse
Immunogen: Mouse gp91[phox] aa. 450-556
Formula Weight [Chemical]: 58kDa
Immunofluorescence, Western Blotting
Specifications
| gp91[phox] | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified |
| 53 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Mouse, Rat | |
| Mouse gp91[phox] aa. 450-556 | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.