missing translation for 'onlineSavingsMsg'
Learn More

Fyn Mouse, Unlabeled, Clone: 25, BD

Mouse Monoclonal Antibody

£223.06 - £441.00

Specifications

Antigen Fyn
Clone 25
Concentration 250μg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15855299
View Documents
BD Biosciences
610163
50 μg
ONLINE EXCLUSIVE PRICE
£233.00
£223.06 / 1 set
Save £9.94
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15865299
View Documents
BD Biosciences
610164
150 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Stimulation of the T cell antigen receptor (TcR) results in protein tyrosine phosphorylation via non-receptor tyrosine kinases. The src-related p59[fyn] kinase is one such enzyme that associates with the TcR. Ligation of the TcR activates the protein kinase activity of p59[fyn] invarious human T cells. Fyn interacts with the CD3-ζ chains through its N-terminal region. In turn, fyn binds other proteins through its SH2 and SH3 domains. These proteins (p82 and p116) may serve as substrates and/or mediators of fyn activity. A 72kDa fyn-related protein binds to the TcR/CD3 complex in certain T cells. This protein, known as p72[fyn-R], is a variant of the fyn kinase and contains at least four novel Ser/Thr phosphorylation sites. These sites may make p72[fyn-R] less susceptible to the action of phosphatases than p59[fyn]. Like p59[fyn], p72[fyn-R] is constitutively associated with its receptor. The exact relationship between these two proteins is not yet clear. However, p72[fyn-R] immunoprecipitates with various antisera raised to either the N- or C-terminus of fyn. Similar to src and yes, fyn is highly
expressed in brain. The localization of fyn in mouse brain and its interaction with myelin-associated glycoprotein (MAG) suggests that the fyn kinase has a role in the sensory network and in myelination during early stages of CNS formation.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

Fyn
250μg/mL
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
IgG2b
Affinity Purified
25
Western Blot
Unconjugated
RUO
Canine, Human, Rat
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Store undiluted at -20°C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.