missing translation for 'onlineSavingsMsg'
Learn More

Fyn Mouse, Unlabeled, Clone: 25, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610163

 View more versions of this product

Product Code. 15855299

  • ONLINE EXCLUSIVE PRICE
    £233.00
    £223.06 / 1 set
    Save £9.94
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Stimulation of the T cell antigen receptor (TcR) results in protein tyrosine phosphorylation via non-receptor tyrosine kinases. The src-related p59[fyn] kinase is one such enzyme that associates with the TcR. Ligation of the TcR activates the protein kinase activity of p59[fyn] invarious human T cells. Fyn interacts with the CD3-ζ chains through its N-terminal region. In turn, fyn binds other proteins through its SH2 and SH3 domains. These proteins (p82 and p116) may serve as substrates and/or mediators of fyn activity. A 72kDa fyn-related protein binds to the TcR/CD3 complex in certain T cells. This protein, known as p72[fyn-R], is a variant of the fyn kinase and contains at least four novel Ser/Thr phosphorylation sites. These sites may make p72[fyn-R] less susceptible to the action of phosphatases than p59[fyn]. Like p59[fyn], p72[fyn-R] is constitutively associated with its receptor. The exact relationship between these two proteins is not yet clear. However, p72[fyn-R] immunoprecipitates with various antisera raised to either the N- or C-terminus of fyn. Similar to src and yes, fyn is highly
expressed in brain. The localization of fyn in mouse brain and its interaction with myelin-associated glycoprotein (MAG) suggests that the fyn kinase has a role in the sensory network and in myelination during early stages of CNS formation.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Fyn
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50 μg
Cell Biology
Canine, Human, Rat
IgG2b
Western Blot
25
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.