missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-FMC63, scFv Antibody, Clone: Y45, Precision Avi, ACROBiosystems
HEK293 Monoclonal Antibody
Brand: ACROBiosystems FM3-BY54-200tests
Description
Specifically recognizes the antigen-recognition domain of FMC63 derived CARs.Specifications
| FMC63 | |
| Monoclonal | |
| Biotin | |
| Human (HEK293) | |
| RUO | |
| Specifically recognizes the antigen-recognition domain of FMC63 derived CARs. | |
| Anti-idiotypic Antibodies | |
| IgG1 κ |
| Flow Cytometry | |
| Y45 | |
| Lyophilized from 0.22μm filtered solution in PBS, 0.5% BSA, pH7.4. Normally trehalose is added as protectant before lyophilization. | |
| 200 Tests | |
| Primary | |
| -20°C | |
| Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction