missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-FMC63, scFv Antibody, Clone: Y45, Precision Avi, ACROBiosystems
HEK293 Monoclonal Antibody
£330.00 - £2065.00
Specifications
| Antigen | FMC63 |
|---|---|
| Clone | Y45 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Biotin |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17256210
|
ACROBiosystems
FM3-BY54-25tests |
25 Tests |
£330.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17246210
|
ACROBiosystems
FM3-BY54-200tests |
200 Tests |
£2065.00
200 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specifically recognizes the antigen-recognition domain of FMC63 derived CARs.Specifications
| FMC63 | |
| Flow Cytometry | |
| Biotin | |
| Human (HEK293) | |
| Lyophilized from 0.22μm filtered solution in PBS, 0.5% BSA, pH7.4. Normally trehalose is added as protectant before lyophilization. | |
| Primary | |
| Specifically recognizes the antigen-recognition domain of FMC63 derived CARs. |
| Y45 | |
| Monoclonal | |
| Powder | |
| RUO | |
| IgG1 κ | |
| -20°C | |
| Anti-idiotypic Antibodies |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title