missing translation for 'onlineSavingsMsg'
Learn More

FAK Mouse, Unlabeled, Clone: 77/FAK, BD

Mouse Monoclonal Antibody

£155.00 - £280.83

Specifications

Antigen FAK
Clone 77/FAK
Concentration 250μg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15865249
View Documents
BD Biosciences
610087
50 μg
£155.00
1 set
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15875249
View Documents
BD Biosciences
610088
150 μg
ONLINE EXCLUSIVE PRICE
£293.00
£280.83 / 1 set
Save £12.17
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Focal Adhesion Kinase (FAK) is a cytoplasmic tyrosine kinase that colocalizes with integrins in focal adhesions. This cellular localization is directed by a 125 amino acid sequence at the C-terminus called the “Focal Adhesion Targeting sequence” (FAT). The binding of extracellular matrix ligands to integrins triggers autophosphorylation and activation of FAK. This creates binding sites for SH2 domains of intracellular signaling molecules such as src, PI3 kinase, and Grb2. FAK's ability to bind numerous structural and signaling proteins via a variety of interactions has led to substantial speculation about its function. Although FAK's precise role has not been elucidated, proposed possibilities include regulating cell motility, cell growth, cytoskeletal organization, and adhesion-dependent cell survival.

Host Species: Mouse
Clone: 77
Isotype: IgG1
Species Reactivity: Human
Immunogen: Chicken FAK aa. 354-533
Formula Weight [Chemical]: 116-125kDa

Bioimaging, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

FAK
250μg/mL
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
IgG1
Affinity Purified
Focal Adhesion Kinase
77/FAK
Western Blot
Unconjugated
RUO
Canine, Chicken, Human, Mouse, Rat
Chicken FAK aa. 354-533
Primary
Store undiluted at -20°C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.