missing translation for 'onlineSavingsMsg'
Learn More

FAK Mouse, Unlabeled, Clone: 77/FAK, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610088

 View more versions of this product

Product Code. 15875249

  • ONLINE EXCLUSIVE PRICE
    £293.00
    £280.83 / 1 set
    Save £12.17
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Focal Adhesion Kinase (FAK) is a cytoplasmic tyrosine kinase that colocalizes with integrins in focal adhesions. This cellular localization is directed by a 125 amino acid sequence at the C-terminus called the “Focal Adhesion Targeting sequence” (FAT). The binding of extracellular matrix ligands to integrins triggers autophosphorylation and activation of FAK. This creates binding sites for SH2 domains of intracellular signaling molecules such as src, PI3 kinase, and Grb2. FAK's ability to bind numerous structural and signaling proteins via a variety of interactions has led to substantial speculation about its function. Although FAK's precise role has not been elucidated, proposed possibilities include regulating cell motility, cell growth, cytoskeletal organization, and adhesion-dependent cell survival.

Host Species: Mouse
Clone: 77
Isotype: IgG1
Species Reactivity: Human
Immunogen: Chicken FAK aa. 354-533
Formula Weight [Chemical]: 116-125kDa

Bioimaging, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

FAK
Monoclonal
250μg/mL
Focal Adhesion Kinase
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Western Blot
77/FAK
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Chicken FAK aa. 354-533
150 μg
Cell Biology
Canine, Chicken, Human, Mouse, Rat
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.