missing translation for 'onlineSavingsMsg'
Learn More

ERK (pan ERK) Mouse, Unlabeled, Clone: 16, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610124

 View more versions of this product

Product Code. 15815279

  • ONLINE EXCLUSIVE PRICE
    £419.00
    £401.97 / 1 set
    Save £17.03
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The family of serine/threonine kinases known as ERKs (extracellular signal regulated kinases) or MAPKs (mitogen-activated protein kinases) are activated after cell stimulation by a variety of hormones and growth factors. Cell stimulation induces a signaling cascade that leads to phosphorylation of MEK (MAPK/ERK kinase) which, in turn, activates ERK via tyrosine and threonine phosphorylation. A myriad of proteins represent the downstream effectors for the active ERK and implicate it in the control of cell proliferation and differentiation, as well as regulation of the cytoskeleton. Activation of ERK is normally transient and cells possess dual specificity phosphatases that are responsible for its down-regulation. Furthermore, multiple studies have shown that elevated ERK activity is associated with some cancers. ERK1 may be observable migrating at 44kDa and ERK2 at 42kDa in addition to a 54kDa ERK and a MAP kinase in the 90kDa range.

Host Species: Mouse
Clone: 16
Isotype: IgG2a
Species Reactivity: Mouse
Immunogen: Rat ERK2 aa. 219-358
Formula Weight [Chemical]: 42-85kDa

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

ERK (pan ERK)
Monoclonal
250μg/mL
Extracellular signal Regulated Kinases
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Western Blot
16
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Rat ERK2 aa. 219-358
150 μg
Cell Biology
Canine, Chicken, Frog, Human, Mouse, Rat
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.