missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Endoglin/CD105 Fluorescein-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 15 publications
£179.00 - £299.00
Specifications
| Antigen | Endoglin/CD105 |
|---|---|
| Clone | 166707 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18718027
![]() |
R&D Systems
FAB10971F-025 |
25 Tests |
£179.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18718396
![]() |
R&D Systems
FAB10971F-100 |
100 Tests |
£299.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Endoglin/CD105 Monoclonal specifically detects Endoglin/CD105 in Human samples. It is validated for Flow Cytometry.Specifications
| Endoglin/CD105 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, ENG, HHT1FLJ41744, ORW, ORW1 | |
| ENG | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human Endoglin/CD105. |
| 166707 | |
| Flow Cytometry | |
| Fluorescein | |
| Mouse | |
| Human | |
| Q5T9B9 | |
| 2022 | |
| Mouse myeloma cell line NS0-derived recombinant human Endoglin/CD105 Glu26-Gly586 Accession # Q5T9B9 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title