missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Endoglin/CD105 Fluorescein-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 15 publications
Brand: R&D Systems FAB10971F-100
This item is not returnable.
View return policy
Description
Endoglin/CD105 Monoclonal specifically detects Endoglin/CD105 in Human samples. It is validated for Flow Cytometry.
Specifications
| Endoglin/CD105 | |
| Monoclonal | |
| Fluorescein | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, ENG, HHT1FLJ41744, ORW, ORW1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2022 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 166707 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Q5T9B9 | |
| ENG | |
| Mouse myeloma cell line NS0-derived recombinant human Endoglin/CD105 Glu26-Gly586 Accession # Q5T9B9 | |
| 100 Tests | |
| Primary | |
| Detects human Endoglin/CD105. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction