missing translation for 'onlineSavingsMsg'
Learn More

Dlg Mouse, Unlabeled, Clone: 12, BD

Mouse Monoclonal Antibody

£148.02 - £428.00

Specifications

Antigen Dlg
Clone 12
Concentration 250μg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15885679
View Documents
BD Biosciences
610874
50 μg
ONLINE EXCLUSIVE PRICE
£155.00
£148.02 / 1 set
Save £6.98
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15895679
View Documents
BD Biosciences
610875
150 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

The human homologue of the Drosophila discs large tumor suppressor protein (hDlg) is a member of the MAGUKs (membrane associated guanylate kinases) protein family. Members of this family (PSD-95, ZO-1, ZO-2, and human erythroid p55) are involved in cell structure and signaling events. The hDlg protein consists of several domains: three PDZ (PSD-95/Discs large/ZO-1) domains, an SH3 domain, and a guanylate kinase-like domain. However, hDlg contains a proline rich N-terminus region consisting of two SH3 domain binding sites that are not normally found in the MAGUKs family. The PDZ domains mediate the interaction of several proteins, such as Shaker-type K+ channel proteins and the APC tumor suppressor protein. Dlg is a peripheral membrane that associates with the cytoskeleton. The cellular location and binding sites of Dlg suggest a role in structure, signal transduction, and growth regulation. Supporting these probable Dlg functions are reports demonstrating that recessive mutations in Drosophila dlg lead to imaginal disc neoplasia and death. Also, Dlg has been reported to bind p56 [lck] tyrosine kinase and the Kv1.3 channel in human T lymphocytes.
Specifications

Specifications

Dlg
250μg/mL
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
IgG1
Affinity Purified
12
Western Blot
Unconjugated
RUO
Human, Mouse, Rat
Human Dlg aa. 5-213
Primary
Store undiluted at -20°C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.