missing translation for 'onlineSavingsMsg'
Learn More

Dlg Mouse, Unlabeled, Clone: 12, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610874

 View more versions of this product

Product Code. 15885679

  • ONLINE EXCLUSIVE PRICE
    £155.00
    £148.02 / 1 set
    Save £6.98
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The human homologue of the Drosophila discs large tumor suppressor protein (hDlg) is a member of the MAGUKs (membrane associated guanylate kinases) protein family. Members of this family (PSD-95, ZO-1, ZO-2, and human erythroid p55) are involved in cell structure and signaling events. The hDlg protein consists of several domains: three PDZ (PSD-95/Discs large/ZO-1) domains, an SH3 domain, and a guanylate kinase-like domain. However, hDlg contains a proline rich N-terminus region consisting of two SH3 domain binding sites that are not normally found in the MAGUKs family. The PDZ domains mediate the interaction of several proteins, such as Shaker-type K+ channel proteins and the APC tumor suppressor protein. Dlg is a peripheral membrane that associates with the cytoskeleton. The cellular location and binding sites of Dlg suggest a role in structure, signal transduction, and growth regulation. Supporting these probable Dlg functions are reports demonstrating that recessive mutations in Drosophila dlg lead to imaginal disc neoplasia and death. Also, Dlg has been reported to bind p56 [lck] tyrosine kinase and the Kv1.3 channel in human T lymphocytes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Dlg
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human Dlg aa. 5-213
50 μg
Cell Biology
Human, Mouse, Rat
IgG1
Western Blot
12
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.