Learn More
Cyclin D3 Mouse, Unlabeled, Clone: 1, BD
Mouse Monoclonal Antibody
£148.02 - £441.00
Specifications
| Antigen | Cyclin D3 |
|---|---|
| Clone | 1 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15855379
|
BD Biosciences
610279 |
50 μg |
£155.00 £148.02 / 1 set Save £6.98 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15865379
|
BD Biosciences
610280 |
150 μg | |||||||
Description
Several classes of cyclins (A-E) have been described. These proteins act as regulatory subunits for cyclin-dependent kinases (cdks). The synthesis and degradation of cyclins is tightly controlled in a cell cycle specific manner. There are at least 3 different D-type cyclins whose relative levels vary among cell types. Cyclin D3 is known to associate with Cdk5 and weakly with Cdk2. Cdk4 is preferentially bound by Cyclin D1. However, in T cells which do not express Cyclin D1, Cdk4 associates with Cyclins D2 and D3. While bound to cdks, the D-type cyclins also associate with the polymerase-δ subunit, PCNA. Cyclin D3 has also been shown to bind pRb in vitro. This suggests that pRb may be an in vivo substrate of D-type cyclins.
This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
| Cyclin D3 | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG2b | |
| Affinity Purified |
| 1 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Canine, Human, Mouse, Rat | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.