Learn More
Cyclin D3 Mouse, Unlabeled, Clone: 1, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 610280
Description
Several classes of cyclins (A-E) have been described. These proteins act as regulatory subunits for cyclin-dependent kinases (cdks). The synthesis and degradation of cyclins is tightly controlled in a cell cycle specific manner. There are at least 3 different D-type cyclins whose relative levels vary among cell types. Cyclin D3 is known to associate with Cdk5 and weakly with Cdk2. Cdk4 is preferentially bound by Cyclin D1. However, in T cells which do not express Cyclin D1, Cdk4 associates with Cyclins D2 and D3. While bound to cdks, the D-type cyclins also associate with the polymerase-δ subunit, PCNA. Cyclin D3 has also been shown to bind pRb in vitro. This suggests that pRb may be an in vivo substrate of D-type cyclins.
This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
| Cyclin D3 | |
| Monoclonal | |
| 250μg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| 150 μg | |
| Cell Biology | |
| Canine, Human, Mouse, Rat | |
| IgG2b |
| Western Blot | |
| 1 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.