Learn More
CRIK Mouse, Unlabeled, Clone: 6, BD
Mouse Monoclonal Antibody
£190.20 - £401.97
Specifications
| Antigen | CRIK |
|---|---|
| Clone | 6 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15895899
|
BD Biosciences
611376 |
50 μg |
£198.00 £190.20 / 1 set Save £7.80 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15805909
|
BD Biosciences
611377 |
150 μg |
£419.00 £401.97 / 1 set Save £17.03 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Citron was identified by its interactions with activated Rho and Rac. It contains a zinc finger (ZF) region, a pleckstrin homology (PH) domain, a citron/MRCK/Gek (CMG) domain, and a coiled-coil domain with a Rho-Rac binding region (RRB). Citron binds to PSD-95 via its C-terminal tSXV motif and the PDZ domain on PSD-95. This association occurs at post synaptic densities where citron-PSD-95 may provide a specific link between the Rho signaling cascade and the synaptic NMDA receptor complex. Citron Rho-interacting kinase (CRIK) and CRIK short kinase (CRIK-SK) are Ser/Thr kinase isoforms of the myotonic dystrophy kinase family. CRIK contains full-length citron, with an N-terminal kinase domain. CRIK-SK is a splice variant that contains only the kinase domain. CRIK co-localizes with actin in keratinocytes and co-expression of CRIK and Rho increases CRIK kinase activity two-fold. In addition, CRIK mutants cause abnormal contraction during cytokinesis. CRIK is expressed highest in testis, but is also present in brain, spleen, lung, and kidney. Thus, CRIK is a Ser/Thr kinase that functions downstream of Rho during events such as cytokinesis and synaptic transmission.
Immunofluorescence, Western Blotting
Specifications
| CRIK | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified | |
| Serine/Threonine Protein Kinase 21 |
| 6 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| Mouse CRIK aa. 1420-1612 | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.